Sodium glycolate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD151466-500GEA 35 EUR
BLDPBD151466-500G BLDPBD151466-100G BLDPBD151466-25G BLDPBD151466-1000G BLDPBD151466-0.025
Sodium glycolate 98%
Sodium glycolate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂H₃NaO₃
MW: 98,03 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00065358
Núm. CAS: 2836-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR