Sodium 2-methyl-2-propanethiolate 98%

BLDPBD151315-5GEA 51 EUR
BLDPBD151315-5G BLDPBD151315-25G BLDPBD151315-1G
Sodium 2-methyl-2-propanethiolate 98%
Sodium 2-methyl-2-propanethiolate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₉NaS
MW: 112,16 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00043311
Núm. CAS: 29364-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR