3,5,7-Trihydroxy-2-(2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)chroman-4-one 98%

BLDPBD151187-5GEA 7 EUR
BLDPBD151187-5G BLDPBD151187-25G BLDPBD151187-100G
3,5,7-Trihydroxy-2-(2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)chroman-4-one 98%
3,5,7-Trihydroxy-2-(2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)chroman-4-one

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₅H₂₂O₁₀
MW: 482,4362 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01075144
Núm. CAS: 42110-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR