Dodecacarbonyltriruthenium 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD150612-250MGEA 29 EUR
BLDPBD150612-250MG BLDPBD150612-1G BLDPBD150612-25G BLDPBD150612-5G
Dodecacarbonyltriruthenium 98%
Dodecacarbonyltriruthenium

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂O₁₂Ru₃
MW: 639,33 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011209
Núm. CAS: 15243-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR