2-Chloro-4'-fluorobenzophenone 96%

Proveedor: BLD PHARMATECH GMBH

BLDPBD15047-5GEA 80 EUR
BLDPBD15047-5G BLDPBD15047-1G BLDPBD15047-250MG
2-Chloro-4'-fluorobenzophenone 96%
2-Chloro-4'-fluorobenzophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₈ClFO
MW: 234,6534 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00000559
Núm. CAS: 1806-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR