N-(4-Bromophenyl)-2,6-dihydroxybenzamide 97%

BLDPBD150305-5GEA 17 EUR
BLDPBD150305-5G BLDPBD150305-25G BLDPBD150305-1G BLDPBD150305-100G
N-(4-Bromophenyl)-2,6-dihydroxybenzamide 97%
N-(4-Bromophenyl)-2,6-dihydroxybenzamide

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₃H₁₀BrNO₃
MW: 308,1274 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00864511
Núm. CAS: 20788-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR