5-Phenyloxazolidin-2-one 97%

BLDPBD150297-1GEA 122 EUR
BLDPBD150297-1G BLDPBD150297-5G BLDPBD150297-250MG
5-Phenyloxazolidin-2-one 97%
5-Phenyloxazolidin-2-one

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₉NO₂
MW: 163,17 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01747962
Núm. CAS: 7693-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR