2,4,6-Trichloro-5-ethylpyrimidine 95%

BLDPBD149830-100MGEA 119 EUR
BLDPBD149830-100MG BLDPBD149830-250MG BLDPBD149830-1G
2,4,6-Trichloro-5-ethylpyrimidine 95%
2,4,6-Trichloro-5-ethylpyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₅Cl₃N₂
MW: 211,4800 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD11112097
Núm. CAS: 1780-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR