3-(Hydrazinylmethyl)pyridine 98%
|
|
BLDPBD149688-100MGEA
19
EUR
BLDPBD149688-100MG
BLDPBD149688-250MG
BLDPBD149688-5G
BLDPBD149688-1G
3-(Hydrazinylmethyl)pyridine 98%
3-(Hydrazinylmethyl)pyridine
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formula:
C₆H₉N₃ MW: 123,16 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD07786307 Núm. CAS: 7112-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...