Potassium (S)-2-hydroxypropanoate 60% in water

BLDPBD148828-1000GEA 66 EUR
BLDPBD148828-1G BLDPBD148828-1000G BLDPBD148828-500G BLDPBD148828-100G BLDPBD148828-25G
Potassium (S)-2-hydroxypropanoate 60% in water
Potassium (S)-2-hydroxypropanoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₃H₅KO₃
MW: 128,1683 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00080809
Núm. CAS: 85895-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR