Polydextrose 95%

BLDPBD148565-25GEA 11 EUR
BLDPBD148565-25G BLDPBD148565-0.025 BLDPBD148565-500G BLDPBD148565-100G
Polydextrose 95%
Polydextrose

Storage condition: Sealed in dry, Room Temperature

Temperatura de almacenaje: Ambiente
Número MDL: MFCD30176547
Núm. CAS: 68424-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR