4-Nitrophenyl butyrate 95%

BLDPBD148443-250MGEA 13 EUR
BLDPBD148443-250MG BLDPBD148443-1G BLDPBD148443-25G BLDPBD148443-5G
4-Nitrophenyl butyrate 95%
4-Nitrophenyl butyrate

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₁₀H₁₁NO₄
MW: 209,1986 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00047734
Núm. CAS: 2635-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR