(2S,3R,4R,5R,6S)-2-Methyl-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol 98%
|
|
BLDPBD148435-1GEA
363
EUR
BLDPBD148435-1G
BLDPBD148435-100MG
BLDPBD148435-250MG
(2S,3R,4R,5R,6S)-2-Methyl-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol 98%
(2S,3R,4R,5R,6S)-2-Methyl-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₂H₁₅NO₇ MW: 285,2500 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00069788 Núm. CAS: 18918-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...