2-(4-((tert-Butoxycarbonyl)amino)-2-nitrophenyl)acetic acid 95%

BLDPBD14804-1GEA 468 EUR
BLDPBD14804-1G BLDPBD14804-250MG BLDPBD14804-100MG
2-(4-((tert-Butoxycarbonyl)amino)-2-nitrophenyl)acetic acid 95%
2-(4-((tert-Butoxycarbonyl)amino)-2-nitrophenyl)acetic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₃H₁₆N₂O₆
MW: 296,2759 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD08236844
Núm. CAS: 512180-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR