3-(Trifluoromethyl)phenylboronic acid pinacol ester 98%
|
|
BLDPBD14769-1GEA
8
EUR
BLDPBD14769-1G
BLDPBD14769-5G
BLDPBD14769-25G
BLDPBD14769-250MG
3-(Trifluoromethyl)phenylboronic acid pinacol ester 98%
3-(Trifluoromethyl)phenylboronic acid pinacol ester
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₃H₁₆BF₃O₂ MW: 272,0712 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD05863925 Núm. CAS: 325142-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...