2-(2,2-Dicyclohexylethyl)piperidine maleate 98%

BLDPBD147311-50MGEA 287 EUR
BLDPBD147311-50MG BLDPBD147311-250MG BLDPBD147311-100MG BLDPBD147311-25MG
2-(2,2-Dicyclohexylethyl)piperidine maleate 98%
2-(2,2-Dicyclohexylethyl)piperidine maleate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₃H₃₉NO₄
MW: 393,5601 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00057329
Núm. CAS: 6724-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR