2-(Benzyloxy)phenylboronic acid 97%

BLDPBD14645-10GEA 53 EUR
BLDPBD14645-10G BLDPBD14645-5G BLDPBD14645-25G BLDPBD14645-1G
2-(Benzyloxy)phenylboronic acid 97%
2-(Benzyloxy)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₃H₁₃BO₃
MW: 228,06 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01632206
Núm. CAS: 190661-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR