4-Benzyloxy-2-fluorophenylboronic acid 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD14610-5GEA 122 EUR
BLDPBD14610-5G BLDPBD14610-25G BLDPBD14610-250MG BLDPBD14610-10G BLDPBD14610-1G BLDPBD14610-0.025G
4-Benzyloxy-2-fluorophenylboronic acid 97%
4-Benzyloxy-2-fluorophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₃H₁₂BFO₃
MW: 246,04 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD02093064
Núm. CAS: 166744-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR