3-Cyano-6-isopropylchromone 98%

BLDPBD14591-1GEA 7 EUR
BLDPBD14591-1G BLDPBD14591-10G BLDPBD14591-25G BLDPBD14591-250MG BLDPBD14591-100G BLDPBD14591-5G
3-Cyano-6-isopropylchromone 98%
3-Cyano-6-isopropylchromone

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₃H₁₁NO₂
MW: 213,2300 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00191961
Núm. CAS: 50743-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR