5-Hydroxy-1-methylimidazolidine-2,4-dione 98%

BLDPBD145857-100MGEA 154 EUR
BLDPBD145857-100MG BLDPBD145857-1G BLDPBD145857-250MG
5-Hydroxy-1-methylimidazolidine-2,4-dione 98%
5-Hydroxy-1-methylimidazolidine-2,4-dione

Storage condition: Inert atmosphere, 2-8°C

Formula: C₄H₆N₂O₃
MW: 130,10 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00899664
Núm. CAS: 84210-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR