Ethyl-4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate 96%
|
|
BLDPBD14509-1GEA
8
EUR
BLDPBD14509-1G
BLDPBD14509-5G
BLDPBD14509-10G
Ethyl-4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate 96%
Ethyl-4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₃H₁₀F₃NO₃ MW: 285,2186 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00173387 Núm. CAS: 23851-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...