N-Isopropyl-2-nitrobenzenesulphonamide 98%
|
|
BLDPBD144821-1GEA
21
EUR
BLDPBD144821-1G
BLDPBD144821-5G
BLDPBD144821-0.025
BLDPBD144821-25G
N-Isopropyl-2-nitrobenzenesulphonamide 98%
N-Isopropyl-2-nitrobenzenesulphonamide
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₁₂N₂O₄S MW: 244,27 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00514604 Núm. CAS: 23530-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...