4-Pyrid-3-ylbenzoic acid 97%

BLDPBD14470-5GEA 125 EUR
BLDPBD14470-5G BLDPBD14470-250MG BLDPBD14470-1G BLDPBD14470-100MG
4-Pyrid-3-ylbenzoic acid 97%
4-Pyrid-3-ylbenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₉NO₂
MW: 199,2054 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03426513
Núm. CAS: 4385-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR