4-(4-Pyridyl)benzaldehyde 98%
|
|
BLDPBD14463-250MGEA
10
EUR
BLDPBD14463-250MG
BLDPBD14463-100MG
BLDPBD14463-10G
BLDPBD14463-0.025G
BLDPBD14463-5G
BLDPBD14463-25G
BLDPBD14463-100G
BLDPBD14463-1G
4-(4-Pyridyl)benzaldehyde 98%
4-(4-Pyridyl)benzaldehyde
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₁₂H₉NO MW: 183,21 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02684110 Núm. CAS: 99163-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...