3-(2-Pyridyl)benzaldehyde 96%

BLDPBD14460-250MGEA 30 EUR
BLDPBD14460-250MG BLDPBD14460-1G BLDPBD14460-10G BLDPBD14460-100MG BLDPBD14460-5G
3-(2-Pyridyl)benzaldehyde 96%
3-(2-Pyridyl)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₉NO
MW: 183,2060 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD02684103
Núm. CAS: 85553-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR