Methyl 2-formamidoacetate 95%

BLDPBD144396-5GEA 45 EUR
BLDPBD144396-5G BLDPBD144396-1G
Methyl 2-formamidoacetate 95%
Methyl 2-formamidoacetate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₄H₇NO₃
MW: 117,10 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00037081
Núm. CAS: 3154-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR