2-Naphthyl boronic acid pinacol ester 97%
|
|
BLDPBD143272-25GEA
93
EUR
BLDPBD143272-25G
BLDPBD143272-1G
BLDPBD143272-250MG
BLDPBD143272-5G
BLDPBD143272-100G
2-Naphthyl boronic acid pinacol ester 97%
2-Naphthyl boronic acid pinacol ester
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₆H₁₉BO₂ MW: 254,14 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD05663868 Núm. CAS: 256652-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...