(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(difluoro(phosphono)methyl)phenyl)propanoic acid 98%

BLDPBD143123-25MGEA 156 EUR
BLDPBD143123-25MG BLDPBD143123-250MG BLDPBD143123-1G BLDPBD143123-100MG BLDPBD143123-50MG
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(difluoro(phosphono)methyl)phenyl)propanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(difluoro(phosphono)methyl)phenyl)propanoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₅H₂₂F₂NO₇P
MW: 517,4153 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01317828
Núm. CAS: 160751-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR