N-acetyl-4-S-cysteaminylphenol 98%

BLDPBD142951-1GEA 140 EUR
BLDPBD142951-1G BLDPBD142951-5G BLDPBD142951-250MG BLDPBD142951-100MG
N-acetyl-4-S-cysteaminylphenol 98%
N-acetyl-4-S-cysteaminylphenol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₃NO₂S
MW: 211,2807 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07779474
Núm. CAS: 91281-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR