N,N-Diethylpiperidine-3-carboxamide 98%

BLDPBD142084-100GEA 891 EUR
BLDPBD142084-100G BLDPBD142084-1G BLDPBD142084-5G BLDPBD142084-25G
N,N-Diethylpiperidine-3-carboxamide 98%
N,N-Diethylpiperidine-3-carboxamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₂₀N₂O
MW: 184,2786 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00005990
Núm. CAS: 3367-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR