2-Ethyl-2-phenylbutanoic acid 96%

BLDPBD14144-5GEA 497 EUR
BLDPBD14144-5G BLDPBD14144-100MG BLDPBD14144-1G BLDPBD14144-250MG
2-Ethyl-2-phenylbutanoic acid 96%
2-Ethyl-2-phenylbutanoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₆O₂
MW: 192,2542 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00075036
Núm. CAS: 5465-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR