N-(2-Mercaptoethyl)acetamide 85%

BLDPBD141027-25GEA 302 EUR
BLDPBD141027-25G BLDPBD141027-1G BLDPBD141027-5G
N-(2-Mercaptoethyl)acetamide 85%
N-(2-Mercaptoethyl)acetamide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₉NOS
MW: 119,18 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00037831
Núm. CAS: 1190-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR