2-((2-Acetamido-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl acetate 97%

BLDPBD14063-5GEA 8 EUR
BLDPBD14063-5G BLDPBD14063-500G BLDPBD14063-1000G BLDPBD14063-25G BLDPBD14063-100G BLDPBD14063-1G
2-((2-Acetamido-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl acetate 97%
2-((2-Acetamido-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl acetate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₁₅N₅O₅
MW: 309,27 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00267706
Núm. CAS: 75128-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR