tert-Butyl 4-amino-4-(2-methoxy-2-oxoethyl)piperidine-1-carboxylate 98%

BLDPBD139424-250MGEA 204 EUR
BLDPBD139424-250MG BLDPBD139424-100MG
tert-Butyl 4-amino-4-(2-methoxy-2-oxoethyl)piperidine-1-carboxylate 98%
tert-Butyl 4-amino-4-(2-methoxy-2-oxoethyl)piperidine-1-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₃H₂₄N₂O₄
MW: 272,3407 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD07784120
Núm. CAS: 362703-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR