Methyl 6-nitropicolinate 95%

BLDPBD139271-250MGEA 170 EUR
BLDPBD139271-250MG BLDPBD139271-100MG BLDPBD139271-1G
Methyl 6-nitropicolinate 95%
Methyl 6-nitropicolinate

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₆N₂O₄
MW: 182,1335 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07776956
Núm. CAS: 26218-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR