Methyl 6-bromoquinoline-2-carboxylate 98%

BLDPBD139234-100MGEA 34 EUR
BLDPBD139234-100MG BLDPBD139234-1G BLDPBD139234-250MG
Methyl 6-bromoquinoline-2-carboxylate 98%
Methyl 6-bromoquinoline-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₈BrNO₂
MW: 266,0900 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08436465
Núm. CAS: 623583-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR