Methyl-5-methoxysalicylate 97%

BLDPBD139176-100GEA 261 EUR
BLDPBD139176-100G BLDPBD139176-5G BLDPBD139176-1G BLDPBD139176-25G
Methyl-5-methoxysalicylate 97%
Methyl-5-methoxysalicylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₉H₁₀O₄
MW: 182,1733 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00017185
Núm. CAS: 2905-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR