Methyl 4-((4-methylpiperazin-1-yl)methyl)benzoate 98%

BLDPBD138856-1GEA 41 EUR
BLDPBD138856-1G BLDPBD138856-250MG BLDPBD138856-25G BLDPBD138856-5G
Methyl 4-((4-methylpiperazin-1-yl)methyl)benzoate 98%
Methyl 4-((4-methylpiperazin-1-yl)methyl)benzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₄H₂₀N₂O₂
MW: 248,32 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01467150
Núm. CAS: 314268-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR