Methyl-3-hydroxy-4-methoxybenzoate 97%

BLDPBD138675-5GEA 15 EUR
BLDPBD138675-5G BLDPBD138675-10G BLDPBD138675-1G BLDPBD138675-500G BLDPBD138675-25G BLDPBD138675-100G
Methyl-3-hydroxy-4-methoxybenzoate 97%
Methyl-3-hydroxy-4-methoxybenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀O₄
MW: 182,1733 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01321262
Núm. CAS: 6702-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR