Methyl-3-amino-4-fluorobenzoate 98%

BLDPBD138574-10GEA 55 EUR
BLDPBD138574-10G BLDPBD138574-25G BLDPBD138574-250MG BLDPBD138574-100G BLDPBD138574-0.025 BLDPBD138574-1G BLDPBD138574-5G
Methyl-3-amino-4-fluorobenzoate 98%
Methyl-3-amino-4-fluorobenzoate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₈FNO₂
MW: 169,1500 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08689743
Núm. CAS: 369-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR