Methyl-2-methyl-4-nitrobenzoate 98%
|
|
BLDPBD138335-25GEA
43
EUR
BLDPBD138335-25G
BLDPBD138335-5G
BLDPBD138335-10G
BLDPBD138335-500G
BLDPBD138335-1G
BLDPBD138335-100G
Methyl-2-methyl-4-nitrobenzoate 98%
Methyl-2-methyl-4-nitrobenzoate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₉NO₄ MW: 195,1700 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03618407 Núm. CAS: 62621-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...