Methyl-2,5-dibromobenzoate 98%
|
|
BLDPBD138097-25GEA
89
EUR
BLDPBD138097-0.025
BLDPBD138097-25G
BLDPBD138097-5G
BLDPBD138097-1G
BLDPBD138097-100G
BLDPBD138097-10G
Methyl-2,5-dibromobenzoate 98%
Methyl-2,5-dibromobenzoate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₆Br₂O₂ MW: 293,94 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00144757 Núm. CAS: 57381-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...