Methyl 1-bromocyclohexanecarboxylate 95%

BLDPBD137930-1GEA 73 EUR
BLDPBD137930-1G BLDPBD137930-250MG BLDPBD137930-5G
Methyl 1-bromocyclohexanecarboxylate 95%
Methyl 1-bromocyclohexanecarboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₁₃BrO₂
MW: 221,0900 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00134214
Núm. CAS: 3196-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR