Methyl 10-bromodecanoate 96%

BLDPBD137893-25GEA 1081 EUR
BLDPBD137893-25G BLDPBD137893-5G BLDPBD137893-250MG BLDPBD137893-1G BLDPBD137893-100MG
Methyl 10-bromodecanoate 96%
Methyl 10-bromodecanoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₂₁BrO₂
MW: 265,1872 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00239436
Núm. CAS: 26825-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR