Linalyl acetate 98%

BLDPBD136351-100GEA 15 EUR
BLDPBD136351-100G BLDPBD136351-25G BLDPBD136351-500G
Linalyl acetate 98%
Linalyl acetate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₂₀O₂
MW: 196,2860 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00008907
Núm. CAS: 115-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR