L(+)-Diisopropyl tartrate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD135658-100GEA 41 EUR
BLDPBD135658-100G BLDPBD135658-25G BLDPBD135658-500G BLDPBD135658-5G BLDPBD135658-0.025
L(+)-Diisopropyl tartrate 98%
L(+)-Diisopropyl tartrate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₈O₆
MW: 234,25 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00064450
Núm. CAS: 2217-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR