Isosorbide dimethyl ether 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD135400-1000GEA 219 EUR
BLDPBD135400-1000G BLDPBD135400-100G BLDPBD135400-500G BLDPBD135400-25G BLDPBD135400-10G BLDPBD135400-5G
Isosorbide dimethyl ether 97%
Isosorbide dimethyl ether

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₄O₄
MW: 174,20 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00064828
Núm. CAS: 5306-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR