(2R,3S)-5-Chloro-2-(((4-methylbenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-methylbenzoate 90%

BLDPBD1353-5GEA 108 EUR
BLDPBD1353-5G BLDPBD1353-0.025G BLDPBD1353-100G BLDPBD1353-250MG BLDPBD1353-10G BLDPBD1353-25G BLDPBD1353-1G
(2R,3S)-5-Chloro-2-(((4-methylbenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-methylbenzoate 90%
(2R,3S)-5-Chloro-2-(((4-methylbenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-methylbenzoate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₂₁H₂₁ClO₅
MW: 388,8400 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00079148
Núm. CAS: 3601-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR