3-Phenyl-4H-chromen-4-one 40%
|
|
BLDPBD135136-100GEA
23
EUR
BLDPBD135136-100G
BLDPBD135136-25G
BLDPBD135136-500G
3-Phenyl-4H-chromen-4-one 40%
3-Phenyl-4H-chromen-4-one
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₅H₁₀O₂ MW: 222,23 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00100851 Núm. CAS: 574-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...