tert-Butyl-4-((methylsulfonyl)oxy)piperidine-1-carboxylate 98%
|
|
BLDPBD13505-100GEA
100
EUR
BLDPBD13505-100G
BLDPBD13505-10G
BLDPBD13505-5G
BLDPBD13505-500G
BLDPBD13505-25G
BLDPBD13505-1G
tert-Butyl-4-((methylsulfonyl)oxy)piperidine-1-carboxylate 98%
tert-Butyl-4-((methylsulphonyl)oxy)piperidine-1-carboxylate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₁₁H₂₁NO₅S MW: 279,3500 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD04116220 Núm. CAS: 141699-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...